Paragon Scientific Cloud Point Standard, Biodiesel
Catalog # 77975-563
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:Cloud point standard, biodiesel
- Size:250 ml
- Storage Temperature:RT
Specifications
About this item
- Shipping conditions: Room temperature