Orders LinkContactUsLinkComponent
CHEMets® Iron Total Ferrous Visual Test
Phenanthroline (total and ferrous)
  77674-810
 :   CHEMetrics
 :  K-6210
CHEMets® Iron Total Ferrous Visual Test
  77674-810
 :  
CHEMets® Iron Total Ferrous Visual Test
CHEMets® Iron Total Ferrous Visual Test
Phenanthroline (total and ferrous)
 :  
$94.64
Each
  • Range: 0 to 1 and 1 to 10 ppm MDL: 0.05 ppm Method: Phenanthroline (total and ferrous)
 : Kit comes in a plastic case and contains everything needed to perform 30 tests:refill, low and high range comparators, activator solution, 25 ml sample cup, and instructions.
  • Chemical method:
    Phenanthroline (total and ferrous)
  • Application:
    Industrial waste water influent and effluent;Industrial process water;Surface and ground water;Sea water;Drinking water