Lipomed Benzoylecgonine-D3.tetrahydrate, 1 mg/ml in Methanol (as free base)
Catalog # 77976-131
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- Description:Benzoylecgonine-D3.tetrahydrate, 1 mg/ml in Methanol (as free base)
- Size:1 ml