Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 3-Chloro-4-(dichloromethyl)-5-hydroxy-2(5H)-furanone
TRC 3-Chloro-4-(dichloromethyl)-5-hydroxy-2(5H)-furanone
TRC 3-Chloro-4-(dichloromethyl)-5-hydroxy-2(5H)-furanone
Catalog # 77979-927
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC 3-Chloro-4-(dichloromethyl)-5-hydroxy-2(5H)-furanone
Catalog # 77979-927
Supplier:
LGC Standards
Supplier Number:
TRC-C365665-2.5MG
New Product
CAS Number:
Specifications
CAS No.:
77439-76-0
Description:
3-Chloro-4-(dichloromethyl)-5-hydroxy-2(5H)-furanone
Size:
2.5 mg
Specifications
About this item
Product Details
Product Details