$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Elagolix Sodium
TRC Elagolix Sodium
77983-790
:
LGC Standards
New Product
:
TRC Elagolix Sodium
77983-790
:
LGC Standards
:
TRC-E501018-100MG
New Product
:
CAS No.:
832720-36-2
Description:
Elagolix Sodium
Size:
100 mg