TRC N-Nitroso-N-methylaniline-d5 (1.0 mg/ml in Methanol)
Catalog # 77987-778
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- CAS No.:1821341-18-7
- Description:N-Nitroso-N-methylaniline-d5 (1.0 mg/ml in Methanol)
- Size:1×1 ml