TRC n-Butyl 4-Hydroxybenzoate-2,3,5,6-d4
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:1219798-67-0
Specifications
Frequently Bought Together
CrossLab Sample Trays for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Degasser Supplies for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Peristaltic Pump Tubing for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
ADM Flow Meter, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Instrument Supplies for GC/MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Tubing Kits for ICP-OES
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Intuvo Flow Chips, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
AVS 6/7 Supplies for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
Supplier: Thermo Scientific
CrossLab Instrument Supplies for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
FRITS REPLACEMENT TWIST 10UM/15UM PK20
Supplier: GLEN RESEARCH CORP MS
O-Rings and Fittings for ICP-OES
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab GC Fittings, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Heat Exchanger for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Fittings for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)