Order Entry
Puerto Rico
ContactUsLinkComponent
 
Lipomed 6-Acetylmorphine.HCl.trihydrate, 1 mg/ml in Methanol (as free base)
  77976-194
New Product
 :  
undefined
Lipomed 6-Acetylmorphine.HCl.trihydrate, 1 mg/ml in Methanol (as free base)
  77976-194
 :  LPM-M-43-HC-1LM
New Product
 :  

 

  • Matrix:
    Methanol
  • CAS No.:
    36418-22-1
  • Description:
    6-Acetylmorphine.HCl.trihydrate, 1 mg/ml in Methanol (as free base)
  • Size:
    1 ml