Paragon Scientific 10 µS/cm @ 25 °C, Conductivity (NIST Traceable, ACS)
Catalog # 77975-547
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:Conductivity standard CRM
- Size:125 ml
- Reference temperature:25 °C
- Certification:ISO 17034
- Conductivity:10 µS/cm