$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Fluticasone Propionate-d3
TRC Fluticasone Propionate-d3
Catalog # 77985-018
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Fluticasone Propionate-d3
Catalog # 77985-018
Supplier:
LGC Standards
Supplier Number:
TRC-F599505-1MG
New Product
CAS Number:
Specifications
Description:
Fluticasone Propionate-d3
Size:
1 mg
Specifications
About this item
Product Details
Product Details