Paragon Scientific 2 M Lithium Chloride Electrolyte, Electrode Filling Solution
Catalog # 77975-848
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Description:2 M Lithium chloride electrolyte, electrode filling solution
- Size:10×30 ml
- Storage Temperature:5 °C
Specifications
About this item
Standard type: Single component
Shipping conditions: Room temperature