$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Ethyl Palmitate
TRC Ethyl Palmitate
Catalog # 77984-435
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Ethyl Palmitate
Catalog # 77984-435
Supplier:
LGC Standards
Supplier Number:
TRC-E925480-500MG
New Product
CAS Number:
Specifications
CAS No.:
628-97-7
Description:
Ethyl Palmitate
Size:
500 mg
Specifications
About this item
Product Details
Product Details