Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC D-Ornithine Hydrochloride
TRC D-Ornithine Hydrochloride
TRC D-Ornithine Hydrochloride
Catalog # 77990-911
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC D-Ornithine Hydrochloride
Catalog # 77990-911
Supplier:
LGC Standards
Supplier Number:
TRC-O695545-1G
New Product
CAS Number:
Specifications
CAS No.:
16682-12-5
Description:
D-Ornithine Hydrochloride
Size:
1 g
Specifications
About this item
Product Details
Product Details