Paragon Scientific RTM2 Mineral Oil Rotational Viscosity
77975-972
New Product
:
- Description:RTM2, Mineral oil rotational viscosity
- Size:500 ml
- Shipping Conditions:RT
- Storage Temperature:RT
- Certification:ISO 17034
- Standard type: Single component