Order Entry
Puerto Rico
ContactUsLinkComponent
CHEMets® Sulfide HR Visual Test Kit (25 to 250 ppm)
CHEMets® Sulfide HR Visual Test Kit (25 to 250 ppm)
Catalog # 77674-870
Supplier:  CHEMetrics
New Product
CAS Number:  
CHEMets® Sulfide HR Visual Test Kit (25 to 250 ppm)
Catalog # 77674-870
Supplier:  CHEMetrics
Supplier Number:  K-9520A
New Product
CAS Number:  

Specifications

  • Chemical method:
    Methylene blue
  • Application:
    Ground water;Waste water;Industrial process water

Specifications

About this item

  • Method: Methylene blue Range: 25 to 250 ppm MDL: 25 ppm
Delivery: The K-9520A Sulfide HR CHEMets® test kit (25 to 250 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): HR CHEMets®refill, flat comparator, 25 ml sample cup, sample cup top, 1.0 ml syringe, sulfide activator solution, and instructions.