$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Dragosantol (>80%)
TRC Dragosantol (>80%)
Catalog # 77983-249
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Dragosantol (>80%)
Catalog # 77983-249
Supplier:
LGC Standards
Supplier Number:
TRC-D480775-1G
New Product
CAS Number:
Specifications
CAS No.:
515-69-5
Description:
Dragosantol (>80%)
Size:
1 g
Specifications
About this item
Product Details
Product Details