Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC O-Acetyl Losartan
TRC O-Acetyl Losartan
TRC O-Acetyl Losartan
77976-613
:
LGC Standards
New Product
:
TRC O-Acetyl Losartan
77976-613
:
LGC Standards
:
TRC-A185300-25MG
New Product
:
CAS No.:
1006062-27-6
Description:
O-Acetyl Losartan
Size:
25 mg