Order Entry
Puerto Rico
ContactUsLinkComponent
Paragon Scientific Polysulfide at 3% (30000 ppm) in Light Mineral Oil
Paragon Scientific Polysulfide at 3% (30000 ppm) in Light Mineral Oil
Catalog # 77975-866
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Paragon Scientific Polysulfide at 3% (30000 ppm) in Light Mineral Oil
Catalog # 77975-866
Supplier:  LGC Standards
Supplier Number:  ALK-LMPS-180
New Product
CAS Number:  

Specifications

  • Description:
    Polysulfide at 3% (30000 ppm) in light mineral oil
  • Size:
    100 ml
  • Storage Temperature:
    RT

Specifications

About this item

  • Shipping conditions: Room temperature