Paragon Scientific Density 25 °C (Nominal: 0.8352 g/ml @ 25 °C)
77975-686
New Product
:
- Pack type:Glass bottle
- Description:Density 25 °C (Nominal: 0.8352 g/ml @ 25 °C), Standard type: Multi-component
- Size:500 ml
- Shipping conditions: Room temperature