Order Entry
Puerto Rico
ContactUsLinkComponent
Lipomed 3,4-Methylenedioxypyrovalerone.HCl, 1 mg/ml in Methanol (as free base)
Lipomed 3,4-Methylenedioxypyrovalerone.HCl, 1 mg/ml in Methanol (as free base)
  77976-239
New Product
 :  
undefined
Lipomed 3,4-Methylenedioxypyrovalerone.HCl, 1 mg/ml in Methanol (as free base)
  77976-239
 :  LPM-PYR-1340-HC-1LM
New Product
 :  

 

  • Matrix:
    Methanol
  • CAS No.:
    24622-62-6
  • Description:
    3,4-Methylenedioxypyrovalerone.HCl, 1 mg/ml in Methanol (as free base)
  • Size:
    1 ml