$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC 4-[(4-Chlorophenyl)methyl]-1(2H)-phthalazinone
TRC 4-[(4-Chlorophenyl)methyl]-1(2H)-phthalazinone
TRC 4-[(4-Chlorophenyl)methyl]-1(2H)-phthalazinone
Supplier:
LGC Standards
New Product
Certificates
Expand image
TRC 4-[(4-Chlorophenyl)methyl]-1(2H)-phthalazinone
Supplier:
LGC Standards
New Product
About this item
Specifications
CAS No.:
53242-88-9
Specifications