$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Lipomed Hydrocodone-D3, 1 mg/ml in Methanol
Lipomed Hydrocodone-D3, 1 mg/ml in Methanol
77976-114
:
LGC Standards
New Product
:
Lipomed Hydrocodone-D3, 1 mg/ml in Methanol
77976-114
:
LGC Standards
:
LPM-C-424-FB-1LM
New Product
:
Matrix:
Methanol
CAS No.:
136765-36-1
Description:
Hydrocodone-D3, 1 mg/ml in Methanol
Size:
1 ml