Lipomed Normeperidine.HCl, 1 mg/ml in Methanol (as free base)
Catalog # 77976-202
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- CAS No.:24465-45-0
- Description:Normeperidine.HCl, 1 mg/ml in Methanol (as free base)
- Size:1 ml