CHEMets® Bromine Visual Test Kit
77674-734
New Product
:
- Chemical method:DPD
- Application:Waste water;Sea water;Food and beverages;Pulp and paper industries
This test uses the DPD testing method and will turn pink in color. It is most often used to measure for accurate dosing of Bromine in water as a disinfectant, but can be used in any applications where low concentrations of bromine in water need to be measured.
- Range: 0 to 2.2 and 0 to11 ppm MDL: 0.125 ppm Method: DPD Analysis time: 1 minute
: The K-1605 bromine CHEMets® test kit comes in a plastic case and contains everything needed to perform 30 tests. CHEMets®refill, flat and round comparators, activator solution, and 25 ml sample cup.