CHEMets® Nitrite Visual Test Kit
77674-834
New Product
:
- Chemical method:Azo dye formation
- Application:Industrial waste water influent and effluent;Industrial process water;Boiler water;Cooling water;Surface and ground water
- Range: 0 to 0.1 and 0 to 1 ppm as N MDL: 0.005 ppm Method: Azo dye formation (NED)
: Kit comes in a plastic case and contains everything needed to perform 30 tests:refill, low and high range comparators, acidifier solution, 25 ml sample cup, and instructions.