Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC N-Desethyl Chloroquine Hydrochloride
TRC N-Desethyl Chloroquine Hydrochloride
TRC N-Desethyl Chloroquine Hydrochloride
77981-732
:
LGC Standards
New Product
:
TRC N-Desethyl Chloroquine Hydrochloride
77981-732
:
LGC Standards
:
TRC-D289505-2.5MG
New Product
:
CAS No.:
4298-11-7
Description:
N-Desethyl Chloroquine Hydrochloride
Size:
2.5 mg