Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 1-(4-Nitrophenyl) 1,4-Benzenedicarboxylate
TRC 1-(4-Nitrophenyl) 1,4-Benzenedicarboxylate
TRC 1-(4-Nitrophenyl) 1,4-Benzenedicarboxylate
77990-652
:
LGC Standards
New Product
:
TRC 1-(4-Nitrophenyl) 1,4-Benzenedicarboxylate
77990-652
:
LGC Standards
:
TRC-N828230-5MG
New Product
:
CAS No.:
3557-99-1
Description:
1-(4-Nitrophenyl) 1,4-Benzenedicarboxylate
Size:
5 mg