Order Entry
Puerto Rico
ContactUsLinkComponent
Film Forming Amines Comparator 2 to 6 ppm
Film Forming Amines Comparator 2 to 6 ppm
Catalog # 77674-586
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Film Forming Amines Comparator 2 to 6 ppm
Catalog # 77674-586
Supplier:  CHEMetrics
Supplier Number:  C-1006
New Product
CAS Number:  

Specifications

  • Chemical method:
    Rose Bengal

Specifications

About this item

The C-1002 Round Filming Amine Comparator is a component of K-1006 Filming Amine CHEMets® Test Kit (Rose Bengal) and is designed to be compared against the color formed in R-1006 Filming Amines CHEMets® Refill (Methyl Orange) ampoules. The kit's range is applicable for routine measurement of residual filming amines in boiler feedwater. The comparator contains a set of color standards (sealed ampoules) containing dye mixtures that are formulated to match the colors produced by the test ampoules. The color standards are pink in color and cover a range of 0 to 1 ppm octadecylamine (ODA). Range: 2 to 6 ppm as ODA or OLA. Increments: 2.0, 3.0, 4.0, 5.0, 6.0 ppm ODA or OLA.