$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Brodifacoum-d4
TRC Brodifacoum-d4
Catalog # 77978-928
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Brodifacoum-d4
Catalog # 77978-928
Supplier:
LGC Standards
Supplier Number:
TRC-B677902-10MG
New Product
CAS Number:
Specifications
Description:
Brodifacoum-d4
Size:
10 mg
Specifications
About this item
Product Details
Product Details