Order Entry
Puerto Rico
ContactUsLinkComponent
Ammonia Vacu-vials® (0 to 3.00 and 0 to 60.0 ppm)
Ammonia Vacu-vials® (0 to 3.00 and 0 to 60.0 ppm)
Catalog # 77674-716
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Ammonia Vacu-vials® (0 to 3.00 and 0 to 60.0 ppm)
Catalog # 77674-716
Supplier:  CHEMetrics
Supplier Number:  K-1413
New Product
CAS Number:  

Specifications

  • Chemical method:
    Hydroxybenzyl Alcohol (HBA)
  • Application:
    Drinking water;Clean surface water;Waste water;Storm water

Specifications

About this item

This kit can be used to determine free ammonia concentration in the presence of monochloramine.

  • Range: 0 to 3.00 and 0 to 60.0 ppm Method: Hydroxybenzyl Alcohol (HBA) Analysis time: 6 minutes
Delivery: The K-1413 ammonia Vacu-vials® test kit (HBA) comes in a cardboard box and contains everything needed to perform 30 tests (except distilled water): 30 Vacu-vials®; ampoules, stabilizer solution, catalyzer solution, activator solution, 25 ml sample cup, 3 ml syringe, ampoule blank, and instructions.