TRC N6-(2-Isopentenyl)adenine
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:2365-40-4
Specifications
Frequently Bought Together
Manual Crimpers and Decappers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® Flat-Bottom Fittings, For Tubing Ø > ¹/₁₆" (1.6 mm) and ≤ ¹/₈" (3.2 mm), IDEX Health & Science
Supplier: Upchurch Scientific
Vaplock™ Chemical Exhaust Filters and Color Indicators, Cole-Parmer
Supplier: Antyila Scientific
Upchurch Scientific® PEEK™ Tubing, IDEX Health & Science
Supplier: Upchurch Scientific
CrossLab Instrument Supplies for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Tubing Kits for ICP-OES
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
InfinityLab Well Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
AVS 6/7 Supplies for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Source Lamps for UV-Vis & UV-Vis-NIR, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Ultra Inert Liners for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Replacement Frits for Empty Cartridges and Empty Cartridges with Pre-installed Frits, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Solvent Bottles and Waste Cans for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Full Flow Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Integrator Supplies and Cables, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
J.T.Baker®, Phase Separation Paper
Supplier: AVANTOR PERFORMANCE MATERIALS US