Paragon Scientific Density 15 °C (Nominal: 0.9413 g/ml @ 15 °C)
77975-651
New Product
:
- Pack type:Glass bottle
- Description:Density 15 °C (Nominal: 0.9413 g/ml @ 15 °C), Standard type: Multi-component
- Size:500 ml
- Shipping conditions: Room temperature