$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC (3R,S)-Oxidosqualene (>90%)
TRC (3R,S)-Oxidosqualene (>90%)
:
LGC Standards
New Product
TRC (3R,S)-Oxidosqualene (>90%)
:
LGC Standards
New Product
CAS No.:
7200-26-2