$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Bodipy C12-Ceramide
TRC Bodipy C12-Ceramide
TRC Bodipy C12-Ceramide
Catalog # 77978-874
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Bodipy C12-Ceramide
Catalog # 77978-874
Supplier:
LGC Standards
Supplier Number:
TRC-B674600-10MG
New Product
CAS Number:
Specifications
CAS No.:
1246355-58-7
Description:
Bodipy C12-Ceramide
Size:
10 mg
Specifications
About this item
Product Details
Product Details