ContactUsLinkComponent
Chlorine Comparator ULR Round 0 to 0.20 ppm
Chlorine Comparator ULR Round 0 to 0.20 ppm
Catalog # 77674-600
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Chlorine Comparator ULR Round 0 to 0.20 ppm
Catalog # 77674-600
Supplier:  CHEMetrics
Supplier Number:  C-2511
New Product
CAS Number:  
Specifications
  • Chemical method:
    DDPD (free and total)
  • Cat. No.:
    77674-600
  • Increments:
    0, 0.04, 0.06, 0.08, 0.10, 0.12, 0.16, 0.20 ppm
  • Range:
    0 - 0.20 ppm
Specifications
About this item
C-2511 Low Range Chlorine Comparator is a component of K-2511 Chlorine (free and total) ULR CHEMets Visual Test Kit and is designed to be compared against the color formed in R-2511 Chlorine ULR CHEMets Refill. The comparator's range is applicable for many disinfection and drinking water applications. Color standards are pink in color and cover a range of 0 to 0.20 ppm chlorine (Cl₂).