TRC 3’-N-Desmethyl-3’-N-tosyl Azithromycin
New Product
- CAS No.:612069-31-5
Frequently Bought Together
CrossLab Sampler and Maintenance Kits for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Headspace Sampler Supplies for GC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Autosampler Valve Supplies for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Fused Silica Tubing Cutters for GC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Tubing Kits for ICP-OES
: AGILENT TECHNOLOGIES, INC (CSD)
Sealing Mats for InfinityLab Well Plates, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Tools for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Sample Loops for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
: Thermo Scientific
AVS 6/7 Supplies for ICP-OES, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab ISIS (Integrated Sample Introduction System) Supplies for ICP-MS (7700/8800), Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Spray Chamber End Caps for ICP-MS, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
FRITS REPLACEMENT TWIST 10UM/15UM PK20
: GLEN RESEARCH CORP MS