Order Entry
Puerto Rico
ContactUsLinkComponent
Lipomed Norhydrocodone.HCl, 1 mg/ml in Acetonitrile/Water : 1/1 (as free base)
Lipomed Norhydrocodone.HCl, 1 mg/ml in Acetonitrile/Water : 1/1 (as free base)
Catalog # 77976-117
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Lipomed Norhydrocodone.HCl, 1 mg/ml in Acetonitrile/Water : 1/1 (as free base)
Catalog # 77976-117
Supplier:  LGC Standards
Supplier Number:  LPM-C-846-HC-1LAH
New Product
CAS Number:  

Specifications

  • Matrix:
    Acetonitrile/Water : 1/1
  • CAS No.:
    71968-04-2
  • Description:
    Norhydrocodone.HCl, 1 mg/ml in Acetonitrile/Water : 1/1 (as free base)
  • Size:
    1 ml

Specifications

About this item