Chromate Instrumental Test 0 to 3.50 ppm
Catalog # 77674-764
Supplier: CHEMetrics
New Product
CAS Number:
Specifications
- Chemical method:Diphenylcarbazide
- Application:Drinking water;Surface and ground water;Domestic and industrial waste water effluent
Specifications
About this item
- Range: 0 to 3.50 ppm Method: Diphenylcarbazide
Delivery: Kit K-2803 comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, acidifier solution, 25 ml sample cup, ampoule blank, and instructions.Vacu to vials kits require the use of the V to 2000 photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately.