CHEMets® Chlorine (Free and Total) Refill (Purpald-Periodate)
Catalog # 77674-890
Supplier: CHEMetrics
New Product
CAS Number:
Specifications
- Chemical method:DDPD (free and total)
- Application:Drinking water;Waste water;Surface and ground water
Specifications
About this item
This product is a refill for K-2511 chlorine (free and total) ULR CHEMets visual kit for the analysis of chlorine in water. It contains 30 self-filling ULR CHEMets ampoules packaged securely in cardboard trays and box. R-2511 chlorine (free and total) - ULR CHEMets refill is compatible with C-2511 chlorine comparator.