$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC 4-Desmethoxypropoxyl-4-methoxy S-Deoxo Rabeprazole
TRC 4-Desmethoxypropoxyl-4-methoxy S-Deoxo Rabeprazole
77981-817
:
LGC Standards
New Product
:
TRC 4-Desmethoxypropoxyl-4-methoxy S-Deoxo Rabeprazole
77981-817
:
LGC Standards
:
TRC-D290615-25MG
New Product
:
CAS No.:
102804-82-0
Description:
4-Desmethoxypropoxyl-4-methoxy S-Deoxo Rabeprazole
Size:
25 mg