Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 4-Methyl Hippuric Acid-d7
TRC 4-Methyl Hippuric Acid-d7
TRC 4-Methyl Hippuric Acid-d7
Catalog # 77989-027
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC 4-Methyl Hippuric Acid-d7
Catalog # 77989-027
Supplier:
LGC Standards
Supplier Number:
TRC-M311977-5MG
New Product
CAS Number:
Specifications
CAS No.:
1216588-60-1
Description:
4-Methyl Hippuric Acid-d7
Size:
5 mg
Specifications
About this item
Product Details
Product Details