Order Entry
Puerto Rico
ContactUsLinkComponent
CHEMets® molybdate Refill 20 to 200 ppm
CHEMets® molybdate Refill 20 to 200 ppm
Catalog # 10065-910
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
CHEMets® molybdate Refill 20 to 200 ppm
Catalog # 10065-910
Supplier:  CHEMetrics
Supplier Number:  R-6720
New Product
CAS Number:  

Specifications

  • Chemical method:
    Catechol
  • Application:
    Industrial water treatment;Power generation;Cooling water systems

Specifications

About this item

This product is a refill for K-6720 molybdate CHEMets visual kit for the analysis of molybdate in water. It contains 30 self-filling CHEMets ampoules packaged securely in cardboard trays and box. R-6720 molybdate CHEMets refill is compatible with C-6720 molybdate comparator.

  • Method: Catechol