Order Entry
Puerto Rico
ContactUsLinkComponent
CHEMets® Hydrogen Peroxide HR Visual Test Kit 5 to 50 ppm
CHEMets® Hydrogen Peroxide HR Visual Test Kit 5 to 50 ppm
Catalog # 77674-794
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
CHEMets® Hydrogen Peroxide HR Visual Test Kit 5 to 50 ppm
Catalog # 77674-794
Supplier:  CHEMetrics
Supplier Number:  K-5520D
New Product
CAS Number:  

Specifications

  • Chemical method:
    Ferric thiocyanate
  • Application:
    Industrial effluent;Waste water;Sea water;Aseptic packaging;Food and beverages

Specifications

About this item

Delivery: The K-5520D hydrogen peroixde HR CHEMets® test kit (5 to 50 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): HR CHEMets®;refill, flat comparator, 25 ml sample cup, 10 ml syringe, and instructions.