$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Levofloxacin-d3 Sodium Salt
TRC Levofloxacin-d3 Sodium Salt
TRC Levofloxacin-d3 Sodium Salt
Catalog # 77987-987
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Levofloxacin-d3 Sodium Salt
Catalog # 77987-987
Supplier:
LGC Standards
Supplier Number:
TRC-L360003-1MG
New Product
CAS Number:
Specifications
CAS No.:
2469039-38-9
Description:
Levofloxacin-d3 Sodium Salt
Size:
1 mg
Specifications
About this item
Product Details
Product Details