Order Entry
Puerto Rico
ContactUsLinkComponent
Paragon Scientific Density 20 °C (Nominal: 1.2800 g/ml @ 20 °C)
Paragon Scientific Density 20 °C (Nominal: 1.2800 g/ml @ 20 °C)
Catalog # 77975-679
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Paragon Scientific Density 20 °C (Nominal: 1.2800 g/ml @ 20 °C)
Catalog # 77975-679
Supplier:  LGC Standards
Supplier Number:  ALK-DEN20-13-500ML
New Product
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Description:
    Density 20 °C (Nominal: 1.2800 g/ml @ 20 °C), Standard type: Multi-component
  • Size:
    500 ml

Specifications

About this item

  • Shipping conditions: Room temperature