Paragon Scientific Sulfur @ 0.005% (50 ppm) in synthetic diesel
Catalog # 77975-734
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Synthetic Diesel
- CAS No.:7704-34-9
- Description:Sulfur @ 0.005% (50 ppm) in synthetic diesel
- Size:100 ml
- Certification:ISO 17034
- Storage Conditions:RT
- Shipping Conditions:RT
- Molecular Weight:32.065