$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Netobimin Sodium Salt
TRC Netobimin Sodium Salt
77990-091
:
LGC Standards
New Product
:
TRC Netobimin Sodium Salt
77990-091
:
LGC Standards
:
TRC-N390900-10MG
New Product
:
CAS No.:
88254-99-3
Description:
Netobimin Sodium Salt
Size:
10 mg