Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 4-Octylphenol
TRC 4-Octylphenol
77990-790
:
LGC Standards
New Product
:
TRC 4-Octylphenol
77990-790
:
LGC Standards
:
TRC-O293775-1G
New Product
:
CAS No.:
1806-26-4
Description:
4-Octylphenol
Size:
1 g