$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Phenoxybenzamine Hydrochloride
TRC Phenoxybenzamine Hydrochloride
TRC Phenoxybenzamine Hydrochloride
Catalog # 77991-349
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Phenoxybenzamine Hydrochloride
Catalog # 77991-349
Supplier:
LGC Standards
Supplier Number:
TRC-P227990-10G
New Product
CAS Number:
Specifications
CAS No.:
63-92-3
Description:
Phenoxybenzamine Hydrochloride
Size:
10 g
Specifications
About this item
Product Details
Product Details