Order Entry
Puerto Rico
ContactUsLinkComponent
Paragon Scientific Type PSU3000, Viscosity Reference Standard
Paragon Scientific Type PSU3000, Viscosity Reference Standard
Catalog # 77975-931
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Paragon Scientific Type PSU3000, Viscosity Reference Standard
Catalog # 77975-931
Supplier:  LGC Standards
Supplier Number:  ALK-PSU3000
New Product
CAS Number:  

Specifications

  • Description:
    Type PSU3000, viscosity reference standard
  • Size:
    500 ml
  • Shipping Conditions:
    RT
  • Storage Temperature:
    RT
  • Certification:
    ISO 17034

Specifications

About this item

  • Standard type: Single component